Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fam213b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025582.3 | NP_079858.2 | 2 | 292 | ||
GenBank mRNA | AK005188.1 | - | 2 | 293 | ||
AK012999.1 | - | 2 | 291 | |||
BC030453.1 | - | 2 | 286 | 5401948 |