Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Uba5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025692.3 | NP_079968.2 | 6 | 697 | ||
GenBank mRNA | AK017787.1 | - | 6 | 697 | ||
AK077825.1 | - | 6 | 678 | |||
AK151823.1 | - | 6 | 654 | |||
AK152029.1 | - | 6 | 692 | |||
AK168907.1 | - | 6 | 691 | |||
AK207610.1 | - | 4 | 261 | |||
BC019764.1 | - | 6 | 574 | 3989119 |