Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CAPRIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005898.4 | NP_005889.3 | 8, 9 | 1072 | ||
NM_203364.2 | NP_976240.1 | 8, 9 | 1072 | |||
XM_017017749.1 | XP_016873238.1 | 7, 8 | 1043 | |||
GenBank mRNA | AK127972.1 | - | 7, 8 | 872 | ||
BC001731.1 | - | 8, 9 | 1072 | 3355481 | ||
Z48042.1 | - | 7, 8 | 925 |