Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lpar6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175116.4 | NP_780325.1 | 1 | 876 | ||
GenBank mRNA | AK008952.1 | - | 1 | 752 | ||
AK011967.1 | - | 1 | 768 | |||
AK032901.1 | - | 1 | 873 | |||
AK169602.1 | - | 1 | 765 | |||
BC047931.1 | - | 1 | 752 | 5251236 | ||
BC057175.1 | - | 1 | 739 | |||
BC069991.1 | - | 1 | 725 | 30244233 | ||
BC145940.1 | - | 1 | 502 | 40131243 | ||
BC145942.1 | - | 1 | 502 | 40131245 |