Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP2 SIRNA Assays ›
Gene Symbol:
MAP2
Gene Name:
microtubule associated protein 2
Gene Aliases:
MAP2A, MAP2B, MAP2C
Chromosome Location:
Chr.2: 209424047 - 209734118 on Build GRCh38
UniGene ID:
Hs.368281
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s8503
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039538.1
NP_001034627.1
11
1539
NM_002374.3
NP_002365.3
12
5233
NM_031845.2
NP_114033.2
9
1165
NM_031847.2
NP_114035.2
9
1165
XM_005246555.1
XP_005246612.1
14
5769
XM_005246565.1
XP_005246622.1
10
5058
XM_005246566.1
XP_005246623.1
10
5046
XM_006712532.1
XP_006712595.1
14
5766
XM_011511186.1
XP_011509488.1
14
5676
XM_011511187.1
XP_011509489.1
13
5570
XM_011511188.1
XP_011509490.1
13
5570
XM_011511190.1
XP_011509492.1
13
5573
XM_011511191.1
XP_011509493.1
12
5438
XM_011511192.1
XP_011509494.1
12
5402
XM_011511193.1
XP_011509495.1
12
5399
XM_011511195.1
XP_011509497.1
12
5324
XM_011511196.2
XP_011509498.1
13
5511
XM_011511197.2
XP_011509499.1
13
5508
XM_017004111.1
XP_016859600.1
12
5399
XM_017004112.1
XP_016859601.1
13
5523
XM_017004113.1
XP_016859602.1
13
5415
XM_017004114.1
XP_016859603.1
12
5352
XM_017004115.1
XP_016859604.1
11
5144
XM_017004116.1
XP_016859605.1
11
5141
XM_017004117.1
XP_016859606.1
11
5141
XM_017004118.1
XP_016859607.1
12
5259
XM_017004119.1
XP_016859608.1
12
5349
XM_017004120.1
XP_016859609.1
12
5349
XM_017004121.1
XP_016859610.1
12
5256
XM_017004122.1
XP_016859611.1
12
5256
XM_017004123.1
XP_016859612.1
12
5340
XM_017004124.1
XP_016859613.1
12
5247
XM_017004125.1
XP_016859614.1
12
5337
XM_017004126.1
XP_016859615.1
12
5337
XM_017004127.1
XP_016859616.1
12
5337
XM_017004128.1
XP_016859617.1
11
5138
XM_017004129.1
XP_016859618.1
11
5135
XM_017004130.1
XP_016859619.1
10
5495
XM_017004131.1
XP_016859620.1
9
4887
XM_017004132.1
XP_016859621.1
9
4875
XM_017004133.1
XP_016859622.1
9
4887
XM_017004134.1
XP_016859623.1
9
4875
XM_017004135.1
XP_016859624.1
9
5321
XM_017004136.1
XP_016859625.1
9
4884
XM_017004137.1
XP_016859626.1
9
4872
XM_017004138.1
XP_016859627.1
10
1362
XM_017004139.1
XP_016859628.1
7
1119
XM_017004140.1
XP_016859629.1
9
1191
GenBank mRNA
AB209330.1
-
8
1083
AK291446.1
-
6
954
AL833567.1
-
3
650
BC027583.1
-
9
1237
BC038857.1
-
11
1528
BC110423.1
-
9
1413
BC117123.1
-
7
994
BC143245.1
-
7
999
S76756.1
-
2
306
U01828.1
-
12
5233
U89329.1
-
6
920
U89330.1
-
9
4976
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008632, NM_001039934, AK166380, AK042880, BC052446, and others.
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
microtubule bundle formation
central nervous system neuron development
neuron projection development
dendrite morphogenesis
dystroglycan binding
structural molecule activity
protein binding
calmodulin binding
microtubule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0