Search
Search

Wdr48| Gene Symbol: Wdr48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026236.3 | NP_080512.1 | 2, 3 | 209 | ||
| GenBank mRNA | AK012599.1 | - | 2, 3 | 209 | ||
| AK018396.1 | - | 2, 3 | 212 | |||
| AK030041.1 | - | 2, 3 | 210 | |||
| AK031244.1 | - | 2, 3 | 222 | |||
| AK033631.1 | - | 2, 3 | 208 | |||
| AK035273.1 | - | 2, 3 | 211 | |||
| AK035864.1 | - | 2, 3 | 211 | |||
| AK043970.1 | - | 2, 3 | 211 | |||
| AK122511.1 | - | 2, 3 | 193 | |||
| AK128953.1 | - | 2, 3 | 209 | |||
| BC006679.1 | - | 1, 2 | 72 | |||
| BC048155.1 | - | 2, 3 | 198 | 5698339 | ||
| BC050106.1 | - | 2, 3 | 226 | |||
| BC062967.1 | - | 2, 3 | 213 | 6835444 | ||