Search
Search

MB| Gene Symbol: MB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005368.2 | NP_005359.1 | 3 | 908 | ||
| NM_203377.1 | NP_976311.1 | 4 | 1000 | |||
| NM_203378.1 | NP_976312.1 | 4 | 983 | |||
| XM_005261605.2 | XP_005261662.1 | 4 | 944 | |||
| GenBank mRNA | AK300657.1 | - | 4 | 866 | ||
| BC014547.1 | - | 3 | 904 | 4244838 | ||
| BC018001.1 | - | 3 | 873 | 4271145 | ||