Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N6amt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159331.1 | NP_001152803.1 | 2, 3 | 300 | ||
NM_026366.2 | NP_080642.1 | 2, 3 | 300 | |||
GenBank mRNA | AK012019.1 | - | 2, 3 | 240 | ||
AK145617.1 | - | 2, 3 | 252 | |||
AK160298.1 | - | 2, 3 | 262 | |||
AY456393.1 | - | 2, 3 | 300 | |||
AY536887.1 | - | 2, 3 | 232 | |||
BC116393.1 | - | 2, 3 | 270 | 40101877 | ||
BC116394.1 | - | 2, 3 | 270 | 40101881 |