Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085377.1 | NP_001078846.1 | 15 | 2664 | ||
NM_002387.2 | NP_002378.1 | 13 | 2208 | |||
XM_005271991.2 | XP_005272048.1 | 13 | 2242 | |||
XM_017009473.1 | XP_016864962.1 | 15 | 2263 | |||
XM_017009474.1 | XP_016864963.1 | 13 | 4928 | |||
GenBank mRNA | AK128596.1 | - | 15 | 2676 | ||
AK300259.1 | - | 13 | 2231 | |||
BC009279.2 | - | 13 | 2208 | 4111706 | ||
BC018919.2 | - | 13 | 2208 | 4131685 | ||
BX537952.1 | - | 13 | 1679 | |||
BX647710.1 | - | 15 | 2138 | |||
M62397.1 | - | 13 | 1898 |