Search
Search

Serpina12| Gene Symbol: Serpina12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026535.2 | NP_080811.1 | 2 | 277 | ||
| GenBank mRNA | AK014346.1 | - | 2 | 219 | ||
| AK014589.1 | - | 2 | 277 | |||
| AY326419.1 | - | 1 | 110 | |||
| BC062143.1 | - | 2 | 231 | 30287005 | ||