Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NDUFA5 SIRNA Assays ›
          Gene Symbol:
          Ndufa5
          Gene Name:
          NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5
          Gene Aliases:
          2900002J19Rik
          Chromosome Location:
          Chr.6: 24518662 - 24527687 on Build GRCm38
          UniGene ID:
          Mm.275780
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s86388

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ndufa5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_026614.2 NP_080890.1 4, 5 398
          GenBank mRNA AK008155.1 - 4, 5 268
          AK008156.1 - 4, 5 264
          AK008286.1 - 4, 5 264
          AK008414.1 - 4, 5 264
          AK009338.1 - 4, 5 282
          AK009396.1 - 4, 5 268
          AK009461.1 - 4, 5 268
          AK009493.1 - 4, 5 267
          AK009608.1 - 4, 5 268
          AK009642.1 - 4, 5 283
          AK009652.1 - 4, 5 268
          AK009775.1 - 4, 5 292
          AK009840.1 - 4, 5 282
          AK009985.1 - 4, 5 282
          AK010024.1 - 4, 5 267
          AK010090.1 - 4, 5 270
          AK010092.1 - 4, 5 284
          AK011579.1 - 4, 5 258
          AK011947.1 - 4, 5 258
          AK019084.1 - 4, 5 269
          AK019201.1 - 4, 5 269
          AK019207.1 - 4, 5 260
          AK019215.1 - 4, 5 268
          AK019242.1 - 4, 5 266
          AK019245.1 - 4, 5 289
          AK019291.1 - 4, 5 269
          AK019301.1 - 4, 5 399
          AK019324.1 - 4, 5 378
          AK019354.1 - 4, 5 271
          AK019359.1 - 4, 5 290
          AK019364.1 - 4, 5 284
          AK019368.1 - 4, 5 282
          AK019430.1 - 4, 5 280
          BC028633.1 - 4, 5 266 1247576

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_012985, BC059148, FQ229037, FQ228773, FQ212236, and others.

          Panther Classification:

          Molecular Function -

          oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          respiratory electron transport chain
          oxidation-reduction process
          NADH dehydrogenase (ubiquinone) activity
          oxidoreductase activity, acting on NAD(P)H

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-slzzl:80/100.66.73.10:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0