Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MECP2 SIRNA Assays ›
          Gene Symbol:
          MECP2
          Gene Name:
          methyl-CpG binding protein 2
          Gene Aliases:
          AUTSX3, MRX16, MRX79, MRXS13, MRXSL, PPMX, RS, RTS, RTT
          Chromosome Location:
          Chr.X: 154021800 - 154097731 on Build GRCh38
          UniGene ID:
          Hs.200716
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s8645
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MECP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001110792.1 NP_001104262.1 3 1488
          NM_001316337.1 NP_001303266.1 5 1780
          NM_004992.3 NP_004983.1 4 1612
          XM_005274682.4 XP_005274739.1 5 1686
          XM_006724819.3 XP_006724882.1 4 1363
          XM_011531166.2 XP_011529468.1 3 1499
          XM_017029533.1 XP_016885022.1 7 2112
          GenBank mRNA AB209464.1 - 3 318
          AF158180.1 - 4 1553
          AJ132917.1 - 3 1461
          AK289444.1 - 4 1587
          AY541280.1 - 3 1429
          BC011612.1 - 3 1470 3956518
          GQ203293.1 - 4 1613
          GQ203294.1 - 4 1612
          GQ203295.1 - 4 1612
          GQ896382.1 - 4 1612
          GU479943.1 - 4 1612
          GU812285.1 - 3 1387
          GU812286.1 - 3 1386
          HM020402.1 - 4 1608
          HM156732.1 - 4 1612
          HM156733.1 - 4 1612
          HQ127345.1 - 4 977
          HQ141377.1 - 3 1386
          HQ141378.1 - 4 1612
          KU178174.1 - 3 1386
          KU178175.1 - 5 1010
          KU178176.1 - 4 927
          KU178177.1 - 4 695
          KU178178.1 - 3 306
          KU178179.1 - 3 304
          KU178180.1 - 3 216
          L37298.1 - 3 1467
          X89430.1 - 2 1357
          X99686.1 - 3 1460
          Y12643.1 - 3 1450

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_022673, XM_006229566, XM_006229567, M94064, DQ897369, and others.

          Matching Taqman Gene Expression Assay:


          Hs00172845_m1

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          behavioral fear response
          response to hypoxia
          startle response
          neurological system process involved in regulation of systemic arterial blood pressure
          regulation of respiratory gaseous exchange by neurological system process
          inositol metabolic process
          mitochondrial electron transport, ubiquinol to cytochrome c
          chromatin silencing
          regulation of gene expression by genetic imprinting
          transcription, DNA-templated
          glutamine metabolic process
          cellular biogenic amine metabolic process
          synapse assembly
          heart development
          respiratory gaseous exchange
          long-term memory
          locomotory behavior
          protein localization
          glucocorticoid metabolic process
          positive regulation of cell proliferation
          adult locomotory behavior
          visual learning
          response to radiation
          pathogenesis
          post-embryonic development
          response to ionizing radiation
          response to lead ion
          dendrite development
          histone methylation
          histone acetylation
          proprioception
          sensory perception of pain
          cerebellum development
          ventricular system development
          negative regulation of histone methylation
          cardiolipin metabolic process
          response to estradiol
          negative regulation of histone acetylation
          social behavior
          neuron maturation
          negative regulation of neuron apoptotic process
          regulation of DNA methylation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          phosphatidylcholine metabolic process
          negative regulation of astrocyte differentiation
          catecholamine secretion
          positive regulation of synapse assembly
          excitatory postsynaptic potential
          long-term synaptic potentiation
          four-way junction DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          mRNA binding
          protein binding
          transcription factor binding
          methyl-CpG binding
          double-stranded methylated DNA binding
          protein domain specific binding
          chromatin DNA binding
          siRNA binding
          histone deacetylase binding
          poly(A) RNA binding
          unmethylated CpG binding
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0