Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pomgnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026651.2 | NP_080927.1 | 2 | 219 | ||
NM_029786.2 | NP_084062.2 | 2 | 219 | |||
GenBank mRNA | AB053220.1 | - | 2 | 201 | ||
AK002638.1 | - | 2 | 211 | |||
BC011201.1 | - | 2 | 142 | 4190368 | ||
BY732188.1 | - | 2 | 219 |