Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ctc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013256.2 | NP_001013274.2 | 11 | 2150 | ||
NM_001143790.1 | NP_001137262.1 | 11 | 2150 | |||
NM_001281464.1 | NP_001268393.1 | 11 | 2021 | |||
NM_001281465.1 | NP_001268394.1 | 11 | 2134 | |||
GenBank mRNA | AK038651.1 | - | 11 | 1937 | ||
AK044055.1 | - | 11 | 1929 | |||
AK145919.1 | - | 11 | 1965 | |||
AK165906.1 | - | 11 | 1950 | |||
AK212869.1 | - | 3 | 339 | |||
BC014687.1 | - | 2 | 321 | |||
BC060629.1 | - | 7 | 1200 | 6847619 | ||
BC066078.1 | - | 11 | 1965 | |||
BC072625.1 | - | 11 | 1978 | |||
BC147495.1 | - | 11 | 1948 | 9056325 | ||
BC147497.1 | - | 11 | 1948 | 9056354 |