Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc38a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027052.3 | NP_081328.2 | 15 | 1609 | ||
GenBank mRNA | AB055004.1 | - | 15 | 1585 | ||
AK005887.1 | - | 2 | 136 | |||
AK051023.1 | - | 15 | 1610 | |||
AK164292.1 | - | 15 | 1552 | |||
AK167539.1 | - | 15 | 1591 | |||
AK169349.1 | - | 14 | 1598 | |||
AY027919.1 | - | 15 | 1648 | |||
BC024072.1 | - | 15 | 1534 | 5097293 | ||
BC024123.1 | - | 15 | 1566 | 5098067 | ||
BC031717.1 | - | 15 | 1527 | 4195569 |