Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253856.1 | NP_001240785.1 | 5 | 461 | ||
NM_001253858.1 | NP_001240787.1 | 4 | 419 | |||
NM_027161.2 | NP_081437.1 | 5 | 479 | |||
GenBank mRNA | AK009779.1 | - | 5 | 442 | ||
BC144992.1 | - | 5 | 478 | 9053540 | ||
BC150685.1 | - | 5 | 459 | 9087596 |