Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kcnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190885.1 | NP_001177814.1 | 6 | 844 | ||
NM_001190886.1 | NP_001177815.1 | 5 | 800 | |||
NM_001290690.1 | NP_001277619.1 | 5 | 999 | |||
NM_027398.3 | NP_081674.2 | 5 | 811 | |||
GenBank mRNA | AB075041.1 | - | 5 | 441 | ||
AJ278534.1 | - | 4 | 369 | |||
AK014313.1 | - | 5 | 800 | |||
AK045948.1 | - | 6 | 845 | |||
AK046398.1 | - | 6 | 833 | |||
AK048178.1 | - | 3 | 2824 | |||
AK081845.1 | - | 5 | 801 | |||
AK082267.1 | - | 4 | 1530 | |||
AY050526.1 | - | 5 | 416 | |||
BC034241.1 | - | 5 | 753 | 4503251 | ||
BU702946.1 | - | 4 | 497 | |||
DQ148493.1 | - | 5 | 753 | |||
DQ148494.1 | - | 5 | 801 |