Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fam135b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177819.3 | NP_808487.2 | 16, 17 | 3811 | ||
GenBank mRNA | AK005816.1 | - | 4, 5 | 679 | ||
BC119174.1 | - | 4, 5 | 427 | |||
BC137579.1 | - | 4, 5 | 427 |