Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc25a33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027460.2 | NP_081736.2 | 3 | 476 | ||
GenBank mRNA | AK017628.1 | - | 3 | 388 | ||
AK157423.1 | - | 3 | 477 | |||
AK187296.1 | - | 2 | 62 | |||
BC011293.1 | - | 3 | 451 | 3989313 | ||
BQ950692.1 | - | 3 | 411 | |||
CK795124.1 | - | 3 | 485 |