Search Thermo Fisher Scientific
Gene Symbol: Nolc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039351.2 | NP_001034440.1 | 11 | 1978 | ||
NM_001039352.2 | NP_001034441.1 | 11 | 1984 | |||
NM_001039353.2 | NP_001034442.1 | 11 | 1981 | |||
NM_053086.3 | NP_444316.2 | 11 | 1981 | |||
GenBank mRNA | AK014336.1 | - | 11 | 1883 | ||
AK029150.1 | - | 11 | 1829 | |||
AK034817.1 | - | 11 | 1858 | |||
AK129040.1 | - | 11 | 1805 | |||
AK147118.1 | - | 11 | 1854 | |||
AK150332.1 | - | 11 | 1815 | |||
AK151031.1 | - | 11 | 1815 | |||
AK152941.1 | - | 11 | 1815 | |||
AK166753.1 | - | 11 | 1887 |