Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MSH2 SIRNA Assays ›
          Gene Symbol:
          MSH2
          Gene Name:
          mutS homolog 2
          Gene Aliases:
          COCA1, FCC1, HNPCC, HNPCC1, LCFS2
          Chromosome Location:
          Chr.2: 47403067 - 47634501 on Build GRCh38
          UniGene ID:
          Hs.597656
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s8966

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MSH2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000251.2 NP_000242.1 10 1691
          NM_001258281.1 NP_001245210.1 11 1507
          XM_005264332.3 XP_005264389.2 10 1638
          XM_011532867.1 XP_011531169.1 10 1638
          GenBank mRNA AK222860.1 - 10 1628
          AK223284.1 - 10 1601
          AK296831.1 - 9 1166
          AK297763.1 - 10 1394
          AK299667.1 - 9 1026
          AK304496.1 - 11 1507
          BC021566.1 - 10 1580 4110354
          BX649122.1 - 10 1586
          DQ648896.1 - 4 378
          L47574.1 - 10 1606
          L47575.1 - 10 1604
          L47576.1 - 10 1605
          L47577.1 - 10 1606
          L47578.1 - 10 1607
          L47579.1 - 10 1606
          L47580.1 - 10 1606
          L47581.1 - 10 1606
          L47582.1 - 10 1606
          L47583.1 - 10 1606
          U03911.1 - 10 1569
          U04045.1 - 10 1634

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          meiotic mismatch repair
          in utero embryonic development
          oxidative phosphorylation
          DNA repair
          mismatch repair
          postreplication repair
          double-strand break repair
          meiotic gene conversion
          cell cycle arrest
          reciprocal meiotic recombination
          germ cell development
          determination of adult lifespan
          male gonad development
          response to X-ray
          response to UV-B
          somatic hypermutation of immunoglobulin genes
          somatic recombination of immunoglobulin gene segments
          B cell mediated immunity
          B cell differentiation
          intra-S DNA damage checkpoint
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          negative regulation of neuron apoptotic process
          maintenance of DNA repeat elements
          negative regulation of reciprocal meiotic recombination
          isotype switching
          negative regulation of DNA recombination
          positive regulation of helicase activity
          magnesium ion binding
          four-way junction DNA binding
          Y-form DNA binding
          heteroduplex DNA loop binding
          double-strand/single-strand DNA junction binding
          DNA binding
          double-stranded DNA binding
          single-stranded DNA binding
          protein binding
          ATP binding
          protein C-terminus binding
          ATPase activity
          centromeric DNA binding
          enzyme binding
          protein kinase binding
          mismatched DNA binding
          guanine/thymine mispair binding
          dinucleotide insertion or deletion binding
          single guanine insertion binding
          single thymine insertion binding
          dinucleotide repeat insertion binding
          oxidized purine DNA binding
          MutLalpha complex binding
          protein homodimerization activity
          ADP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0