Search Thermo Fisher Scientific
Gene Symbol: MSX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002448.3 | NP_002439.2 | 2 | 1362 | ||
GenBank mRNA | AK290636.1 | - | 2 | 1363 | ||
BC021285.2 | - | 2 | 1329 | 5018103 | ||
BC067353.1 | - | 2 | 1122 | 4893288 | ||
BU624994.1 | - | 304 | ||||
M97676.1 | - | 2 | 1348 |