Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DUS4L SIRNA Assays ›
Gene Symbol:
Dus4l
Gene Name:
dihydrouridine synthase 4-like (S. cerevisiae)
Gene Aliases:
2310069P03Rik, 2700089B10Rik, AI482040, Pp35
Chromosome Location:
Chr.12: 31640050 - 31654826 on Build GRCm38
UniGene ID:
Mm.62979
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s90298
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Dus4l
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_028002.2
NP_082278.1
5
560
GenBank mRNA
AK010138.1
-
5
561
BC109351.2
-
5
495
40056894
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001135803, XM_006239990, XM_006239991, XM_006239992, XM_006239993, and others.
Panther Classification:
Molecular Function -
RNA processing factor
Gene Ontology Categories:
Function(s)
Process(es)
tRNA processing
oxidation-reduction process
catalytic activity
oxidoreductase activity
enoyl-[acyl-carrier-protein] reductase activity
tRNA dihydrouridine synthase activity
2,3-dihydroxy-2,3-dihydro-phenylpropionate dehydrogenase activity
cis-2,3-dihydrodiol DDT dehydrogenase activity
trans-9R,10R-dihydrodiolphenanthrene dehydrogenase activity
cis-chlorobenzene dihydrodiol dehydrogenase activity
2,5-dichloro-2,5-cyclohexadiene-1,4-diol dehydrogenase activity
trans-1,2-dihydrodiolphenanthrene dehydrogenase activity
3,4-dihydroxy-3,4-dihydrofluorene dehydrogenase activity
benzo(a)pyrene-trans-11,12-dihydrodiol dehydrogenase activity
benzo(a)pyrene-cis-4,5-dihydrodiol dehydrogenase activity
citronellyl-CoA dehydrogenase activity
menthone dehydrogenase activity
phthalate 3,4-cis-dihydrodiol dehydrogenase activity
cinnamate reductase activity
flavin adenine dinucleotide binding
NADPH-dependent curcumin reductase activity
NADPH-dependent dihydrocurcumin reductase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0