Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fbxo22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028049.2 | NP_082325.2 | 7 | 1223 | ||
GenBank mRNA | AK002745.1 | - | 7 | 1103 | ||
AK005473.1 | - | 6 | 1051 | |||
BC018273.1 | - | 7 | 1163 | 4458296 |