Search
Search

MTNR1A| Gene Symbol: MTNR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005958.4 | NP_005949.1 | 2 | 914 | ||
| XM_011532002.2 | XP_011530304.1 | 2 | 995 | |||
| GenBank mRNA | AF435588.1 | - | 1 | 321 | ||
| BC074946.2 | - | 2 | 730 | 30915497 | ||
| BC074947.2 | - | 2 | 730 | 30915274 | ||
| BC113816.1 | - | 1 | 365 | |||
| BC126297.1 | - | 2 | 914 | 8992013 | ||
| BC126299.1 | - | 2 | 912 | 8992015 | ||
| EU432127.1 | - | 2 | 712 | |||
| U14108.1 | - | 2 | 744 | |||
Set Membership: |
|