Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Chst14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028117.3 | NP_082393.3 | 1 | 1211 | ||
GenBank mRNA | AK011230.1 | - | 1 | 1148 | ||
AK154555.1 | - | 1 | 1200 | |||
AK154636.1 | - | 1 | 1180 | |||
AK155115.1 | - | 2 | 1119 | |||
AK159357.1 | - | 1 | 1179 | |||
AK171614.1 | - | 1 | 1081 | |||
AK208059.1 | - | 1 | 43 | |||
BC026886.1 | - | 1 | 1032 | |||
BC043700.1 | - | 1 | 1184 | 5346204 | ||
BC085479.1 | - | 1 | 1128 | 6415106 |