Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wdsub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159636.1 | NP_001153108.1 | 6 | 1011 | ||
NM_028118.2 | NP_082394.1 | 6 | 1011 | |||
GenBank mRNA | AK011391.1 | - | 6 | 947 | ||
BC050792.1 | - | 6 | 952 | |||
BC061948.1 | - | 6 | 989 | 6312489 |