Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Acbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145781.1 | NP_001139253.1 | 3 | 863 | ||
NM_026683.1 | NP_080959.1 | 3 | 863 | |||
NM_028250.3 | NP_082526.2 | 3 | 863 | |||
GenBank mRNA | AK002470.1 | - | 3 | 864 | ||
AK011781.1 | - | 3 | 856 | |||
AK082937.1 | - | 3 | 864 | |||
AK165836.1 | - | 3 | 858 | |||
AK167959.1 | - | 4 | 804 | |||
AK209754.1 | - | 3 | 178 | |||
BC028537.1 | - | 3 | 608 | |||
BC061029.1 | - | 3 | 589 | |||
BC115580.1 | - | 3 | 455 | 40102241 | ||
BC115581.1 | - | 3 | 455 | 40102242 |