Search
Search

Atxn7l2| Gene Symbol: Atxn7l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289545.1 | NP_001276474.1 | 6 | 913 | ||
| NM_175183.5 | NP_780392.1 | 7 | 1092 | |||
| GenBank mRNA | AK046849.1 | - | 7 | 1500 | ||
| AK131184.1 | - | 1 | 759 | |||
| BC027136.1 | - | 3 | 1390 | |||
| BC088996.1 | - | 5 | 687 | |||
| BC094547.1 | - | 6 | 848 | |||
| BC137599.1 | - | 6 | 834 | 8860618 | ||
| BC137600.1 | - | 6 | 861 | 8860619 | ||
| BC144756.1 | - | 6 | 841 | 9053304 | ||