Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hspa12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028306.3 | NP_082582.1 | 11 | 1272 | ||
GenBank mRNA | AK012548.1 | - | 11 | 1245 | ||
AY196790.1 | - | 10 | 1152 | |||
BC011103.1 | - | 11 | 1272 | 3990088 |