Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bmper | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028472.2 | NP_082748.1 | 13 | 2073 | ||
GenBank mRNA | AF454954.1 | - | 13 | 2073 | ||
AK014221.1 | - | 11 | 1278 | |||
AY263358.1 | - | 13 | 1594 | |||
BC042718.1 | - | 1 | 83 | |||
BC066153.1 | - | 13 | 1578 | 30098453 |