Search Thermo Fisher Scientific
Gene Symbol: MYH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002472.2 | NP_002463.2 | 28, 29 | 3964 | ||
GenBank mRNA | M36769.1 | - | 27, 28 | 3942 | ||
X51592.1 | - | 14, 15 | 2369 | |||
Z38133.1 | - | 27, 28 | 3942 |