Search
Search

Phactr3| Gene Symbol: Phactr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007154.3 | NP_001007155.1 | 10, 11 | 1433 | ||
| NM_001177789.1 | NP_001171260.1 | 10, 11 | 1651 | |||
| NM_001177790.1 | NP_001171261.1 | 10, 11 | 1648 | |||
| NM_001177791.1 | NP_001171262.1 | 10, 11 | 1485 | |||
| NM_028806.2 | NP_083082.1 | 10, 11 | 1890 | |||
| GenBank mRNA | AJ632220.1 | - | 10, 11 | 1448 | ||
| AJ632221.1 | - | 10, 11 | 1486 | |||
| AJ632222.1 | - | 10, 11 | 1433 | |||
| AJ632223.1 | - | 9, 10 | 1323 | |||
| AK005136.1 | - | 10, 11 | 1455 | |||
| AK032242.1 | - | 10, 11 | 1657 | |||
| AK038449.1 | - | 10, 11 | 1401 | |||
| AK135144.1 | - | 10, 11 | 1891 | |||
| AK220554.1 | - | 10, 11 | 1648 | |||
| AY501387.1 | - | 10, 11 | 1597 | |||
| BC058621.1 | - | 10, 11 | 1806 | 5708186 | ||
| BC059869.1 | - | 10, 11 | 1669 | 6809310 | ||