Search
Search

Chpf| Gene Symbol: Chpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001565.2 | NP_001001565.1 | 5 | 2364 | ||
| NM_001001566.3 | NP_001001566.1 | 4 | 2221 | |||
| GenBank mRNA | AK006466.1 | - | 4 | 1592 | ||
| AK155424.1 | - | 4 | 1882 | |||
| AK160734.1 | - | 3 | 2469 | |||
| BC030889.1 | - | 4 | 1709 | |||
| BC057050.1 | - | 5 | 2200 | |||
| BC060598.1 | - | 5 | 2200 | |||
| BC071273.1 | - | 4 | 2021 | 6334418 | ||