Search
Search

Cpa5| Gene Symbol: Cpa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144537.3 | NP_653120.1 | 8, 9 | 1276 | ||
| GenBank mRNA | AF466283.1 | - | 8, 9 | 1277 | ||
| AK015256.1 | - | 8, 9 | 1278 | |||
| BC138114.1 | - | 7, 8 | 794 | 8861133 | ||
| BC145151.1 | - | 7, 8 | 794 | |||