Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Uhrf1bp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029166.2 | NP_083442.2 | 6 | 871 | ||
GenBank mRNA | AB093261.1 | - | 6 | 836 | ||
AK033300.1 | - | 7 | 879 | |||
AK087280.1 | - | 4 | 423 | |||
AK141050.1 | - | 6 | 880 | |||
BB529788.2 | - | 2 | 263 | |||
BC030383.1 | - | 830 | ||||
BC132134.1 | - | 6 | 664 | 40130411 | ||
BC138092.1 | - | 6 | 664 | 8861111 |