Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NDUFA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005002.4 | NP_004993.1 | 3 | 270 | ||
GenBank mRNA | AF050641.1 | - | 3 | 260 | ||
AK291067.1 | - | 3 | 247 | |||
AK308588.1 | - | 3 | 251 | |||
BC003351.1 | - | 2 | 124 | |||
BC009311.2 | - | 3 | 251 | 4106931 | ||
BC015837.1 | - | 3 | 266 | 4293690 | ||
BC111546.1 | - | 3 | 242 | 5761015 | ||
CB110957.1 | - | 3 | 270 | |||
L04490.1 | - | 3 | 236 |