Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NDUFB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168331.1 | NP_001161803.1 | 2 | 312 | ||
NM_004547.5 | NP_004538.2 | 2 | 312 | |||
GenBank mRNA | AF044957.1 | - | 2 | 241 | ||
BC000855.1 | - | 2 | 284 | 3459215 | ||
BC070285.1 | - | 2 | 251 | 6714676 | ||
BC146885.1 | - | 2 | 283 | 9056813 | ||
BC146887.1 | - | 2 | 283 | 9056815 | ||
BC146927.1 | - | 2 | 283 | 9056855 | ||
BC146932.1 | - | 2 | 283 | 9056860 | ||
BF212059.1 | - | 311 | ||||
BF978138.1 | - | 312 |