Search
Search

Fggy| Gene Symbol: Fggy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113412.1 | NP_001106883.1 | 9, 10 | 1111 | ||
| NM_029347.2 | NP_083623.1 | 9, 10 | 1160 | |||
| GenBank mRNA | AK009249.1 | - | 3, 4 | 281 | ||
| AK082737.1 | - | 6, 7 | 691 | |||
| AK085534.1 | - | 9, 10 | 1122 | |||
| AK138439.1 | - | 9, 10 | 1108 | |||
| BC031708.1 | - | 9, 10 | 1078 | 4194716 | ||