Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prl7b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029355.2 | NP_083631.1 | 5, 6 | 602 | ||
GenBank mRNA | AK005458.1 | - | 5, 6 | 603 | ||
BC116772.1 | - | 5, 6 | 579 | 40126091 | ||
BC119183.1 | - | 5, 6 | 579 | 8733932 |