Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cul5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161618.1 | NP_001155090.1 | 7 | 1221 | ||
NM_027807.3 | NP_082083.2 | 8 | 1302 | |||
GenBank mRNA | AK014894.1 | - | 8 | 895 | ||
AK031955.1 | - | 8 | 1305 | |||
AK034293.1 | - | 5 | 592 | |||
AK046030.1 | - | 8 | 1273 | |||
AK162082.1 | - | 1225 | ||||
BC075710.1 | - | 8 | 1231 | 6312416 | ||
CV559336.1 | - | 4 | 518 |