Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mff | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029409.2 | NP_083685.2 | 3, 4 | 491 | ||
GenBank mRNA | AK017226.1 | - | 3, 4 | 491 | ||
AK083056.1 | - | 3, 4 | 476 | |||
AK139649.1 | - | 3, 4 | 470 | |||
AK177568.1 | - | 1, 2 | 105 | |||
AK178118.1 | - | 1, 2 | 105 | |||
AK180892.1 | - | 1, 2 | 138 | |||
AK181587.1 | - | 1, 2 | 105 | |||
AK183556.1 | - | 1, 2 | 105 | |||
AK188151.1 | - | 1, 2 | 88 | |||
AK199555.1 | - | 1, 2 | 105 | |||
AK207673.1 | - | 1, 2 | 105 | |||
AK207808.1 | - | 1, 2 | 55 | |||
AK209807.1 | - | 1, 2 | 105 | |||
AK210863.1 | - | 1, 2 | 105 | |||
AK212140.1 | - | 1, 2 | 62 | |||
AK212284.1 | - | 1, 2 | 105 | |||
AK212704.1 | - | 1, 2 | 105 | |||
AK213477.1 | - | 1, 2 | 105 | |||
AK214594.1 | - | 1, 2 | 74 | |||
AK214622.1 | - | 1, 2 | 105 | |||
AK215829.1 | - | 1, 2 | 50 | |||
AK216481.1 | - | 1, 2 | 105 | |||
AK216685.1 | - | 1, 2 | 55 | |||
AK218828.1 | - | 1, 2 | 105 | |||
AK218934.1 | - | 3, 4 | 348 | |||
AK219490.1 | - | 1, 2 | 105 | |||
BC016597.1 | - | 1, 2 | 202 | 3489844 | ||
BC059229.1 | - | 3, 4 | 491 | 6413193 |