Search
Search

Ulbp1| Gene Symbol: Ulbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029975.2 | NP_084251.1 | 6 | 1054 | ||
| GenBank mRNA | AK020784.1 | - | 6 | 929 | ||
| AK084712.1 | - | 1153 | ||||
| AK085119.1 | - | 1216 | ||||
| AY249021.1 | - | 5 | 683 | |||
| BC104292.1 | - | 3 | 301 | |||
| BC104293.1 | - | 3 | 301 | |||
| BC114390.1 | - | 3 | 300 | |||
| BC114420.1 | - | 3 | 300 | |||
| BC127022.1 | - | 3 | 301 | |||
| BC127023.1 | - | 3 | 301 | |||
| BC132070.1 | - | 6 | 1054 | 40130347 | ||
| BC132072.1 | - | 6 | 1054 | 40130349 | ||