Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
CDMO and Clinical Trial Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NFYC SIRNA Assays ›
Gene Symbol:
NFYC
Gene Name:
nuclear transcription factor Y subunit gamma
Gene Aliases:
CBF-C, CBFC, H1TF2A, HAP5, HSM, NF-YC
Chromosome Location:
Chr.1: 40691570 - 40771603 on Build GRCh38
UniGene ID:
Hs.713051
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s9534
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NFYC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142587.1
NP_001136059.1
2, 3
406
NM_001142588.1
NP_001136060.1
2, 3
406
NM_001142589.1
NP_001136061.1
2, 3
406
NM_001142590.1
NP_001136062.1
7, 8
783
NM_001308114.1
NP_001295043.1
1, 2
108
NM_001308115.1
NP_001295044.1
2, 3
402
NM_014223.4
NP_055038.2
2, 3
406
XM_005270894.1
XP_005270951.1
2, 3
402
XM_005270895.3
XP_005270952.3
2, 3
258
XM_006710658.2
XP_006710721.2
2, 3
258
XM_006710660.2
XP_006710723.2
2, 3
258
XM_006710661.1
XP_006710724.1
2, 3
287
XM_006710662.2
XP_006710725.2
2, 3
258
XM_011541516.1
XP_011539818.1
2, 3
207
XM_011541517.1
XP_011539819.1
2, 3
258
XM_017001364.1
XP_016856853.1
2, 3
427
XM_017001365.1
XP_016856854.1
2, 3
287
XM_017001366.1
XP_016856855.1
2, 3
361
XM_017001367.1
XP_016856856.1
2, 3
402
XM_017001368.1
XP_016856857.1
2, 3
402
XM_017001369.1
XP_016856858.1
3, 4
759
GenBank mRNA
AB208975.1
-
2, 3
233
AF191744.1
-
1, 2
107
AK000346.1
-
2, 3
359
AK127677.1
-
7, 8
778
AK300774.1
-
2, 3
209
AK301385.1
-
2, 3
290
BC005003.1
-
2, 3
255
3502903
BP230898.1
-
2, 3
402
BT020081.1
-
1, 2
100
D85425.1
-
2, 3
183
D89986.1
-
2, 3
168
DC297902.1
-
4, 5
518
DC391720.1
-
2, 3
406
DC428152.1
-
2, 3
406
U62296.1
-
1, 2
100
U78774.1
-
3, 4
274
Z74792.1
-
2, 3
285
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein folding
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
protein binding
transcription factor binding
transcription regulatory region DNA binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-lhtlm:80/10.239.167.245:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0