Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271725.1 | NP_001258654.1 | 2 | 277 | ||
NM_001271726.1 | NP_001258655.1 | 2 | 313 | |||
NM_001271727.1 | NP_001258656.1 | 2 | 582 | |||
NM_001271728.1 | NP_001258657.1 | 2 | 380 | |||
NM_030706.3 | NP_109631.2 | 2 | 376 | |||
GenBank mRNA | AB043550.1 | - | 2 | 360 | ||
AF220017.1 | - | 2 | 225 | |||
AK039741.1 | - | 2 | 374 | |||
AK045410.1 | - | 2 | 373 | |||
AK046887.1 | - | 2 | 361 | |||
AK079415.1 | - | 2 | 527 | |||
AK083057.1 | - | 2 | 374 | |||
AK141419.1 | - | 2 | 223 | |||
AK147275.1 | - | 2 | 490 | |||
AK147395.1 | - | 2 | 371 | |||
AK164350.1 | - | 1 | 79 | |||
AK220343.1 | - | 2 | 581 | |||
BC058668.1 | - | 2 | 344 | |||
BC058961.1 | - | 2 | 380 | 6826143 | ||
CD802800.1 | - | 2 | 239 | |||
CD803806.1 | - | 2 | 376 | |||
CJ100776.1 | - | 2 | 278 |