Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmprss5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030709.2 | NP_109634.2 | 7 | 689 | ||
GenBank mRNA | AB016229.1 | - | 7 | 687 | ||
AB016230.1 | - | 7 | 689 | |||
AB016423.1 | - | 3 | 304 | |||
AB041037.1 | - | 7 | 646 | |||
AK029714.1 | - | 8 | 1536 | |||
BC132151.1 | - | 6 | 635 | 40130428 | ||
BC145027.1 | - | 6 | 617 | 9053575 |