Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000620.4 | NP_000611.1 | 13 | 2866 | ||
NM_001204213.1 | NP_001191142.1 | 12 | 1458 | |||
NM_001204214.1 | NP_001191143.1 | 12 | 1453 | |||
NM_001204218.1 | NP_001191147.1 | 13 | 2866 | |||
XM_011538398.2 | XP_011536700.1 | 14 | 2737 | |||
XM_017019345.1 | XP_016874834.1 | 13 | 2873 | |||
XM_017019346.1 | XP_016874835.1 | 13 | 2687 | |||
XM_017019347.1 | XP_016874836.1 | 13 | 2776 | |||
GenBank mRNA | AK294435.1 | - | 13 | 2670 | ||
AK307481.1 | - | 13 | 2866 | |||
D16408.1 | - | 12 | 2155 | |||
EU753241.1 | - | 13 | 2840 | |||
L02881.1 | - | 12 | 2582 | |||
U17327.1 | - | 13 | 2840 | |||
U31466.1 | - | 12 | 2162 |