Search
Search

NOS1| Gene Symbol: NOS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000620.4 | NP_000611.1 | 17 | 3281 | ||
| NM_001204213.1 | NP_001191142.1 | 16 | 1873 | |||
| NM_001204214.1 | NP_001191143.1 | 16 | 1868 | |||
| NM_001204218.1 | NP_001191147.1 | 18 | 3383 | |||
| XM_011538398.2 | XP_011536700.1 | 19 | 3254 | |||
| XM_017019345.1 | XP_016874834.1 | 18 | 3390 | |||
| XM_017019346.1 | XP_016874835.1 | 18 | 3204 | |||
| XM_017019347.1 | XP_016874836.1 | 17 | 3191 | |||
| GenBank mRNA | AJ004918.1 | - | 3 | 170 | ||
| AK294435.1 | - | 17 | 3085 | |||
| AK307481.1 | - | 17 | 3281 | |||
| D16408.1 | - | 16 | 2570 | |||
| EU753241.1 | - | 17 | 3255 | |||
| L02881.1 | - | 16 | 2997 | |||
| U17327.1 | - | 17 | 3255 | |||
| U31466.1 | - | 16 | 2577 | |||