Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NPC1 SIRNA Assays ›
          Gene Symbol:
          NPC1
          Gene Name:
          NPC intracellular cholesterol transporter 1
          Gene Aliases:
          NPC
          Chromosome Location:
          Chr.18: 23506184 - 23586617 on Build GRCh38
          UniGene ID:
          Hs.464779
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s9667

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NPC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000271.4 NP_000262.2 23, 24 3873
          XM_005258277.1 XP_005258334.1 23, 24 3813
          XM_005258278.4 XP_005258335.1 23, 24 3813
          XM_005258279.2 XP_005258336.1 23, 24 3762
          XM_006722479.3 XP_006722542.1 23, 24 3813
          XM_011526015.1 XP_011524317.1 23, 24 4061
          XM_017025784.1 XP_016881273.1 23, 24 3813
          XM_017025785.1 XP_016881274.1 23, 24 3813
          XM_017025786.1 XP_016881275.1 23, 24 3762
          XM_017025787.1 XP_016881276.1 23, 24 3762
          GenBank mRNA AB209048.1 - 23, 24 3634
          AB971140.1 - 23, 24 3599
          AF002020.1 - 23, 24 3722
          AK293779.1 - 16, 17 2677
          BC063302.1 - 23, 24 3696 30340517

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          endocytosis
          autophagy
          lysosomal transport
          signal transduction
          adult walking behavior
          cholesterol metabolic process
          bile acid metabolic process
          negative regulation of macroautophagy
          cholesterol transport
          membrane raft organization
          cholesterol efflux
          response to drug
          cholesterol homeostasis
          response to cadmium ion
          viral entry into host cell
          negative regulation of cell death
          cellular response to steroid hormone stimulus
          cellular response to low-density lipoprotein particle stimulus
          establishment of protein localization to membrane
          positive regulation of cholesterol homeostasis
          virus receptor activity
          receptor activity
          transmembrane signaling receptor activity
          protein binding
          sterol transporter activity
          cholesterol binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nq57x:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0