Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC4A10 SIRNA Assays ›
          Gene Symbol:
          Slc4a10
          Gene Name:
          solute carrier family 4, sodium bicarbonate cotransporter-like, member 10
          Gene Aliases:
          NCBE, mKIAA4136
          Chromosome Location:
          Chr.2: 62046515 - 62326743 on Build GRCm38
          UniGene ID:
          Mm.314497
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s97071

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Slc4a10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242378.1 NP_001229307.1 11 1434
          NM_001242379.1 NP_001229308.1 10 1344
          NM_001242380.1 NP_001229309.1 11 1434
          NM_001242381.1 NP_001229310.1 10 1341
          NM_001242382.1 NP_001229311.1 11 1431
          NM_001242383.1 NP_001229312.1 10 1341
          NM_033552.3 NP_291030.2 10 1344
          GenBank mRNA AB033759.1 - 10 1265
          AK043017.1 - 10 1320
          AK220501.1 - 11 1399
          BC039226.1 - 10 1293 4480841
          HM149278.1 - 10 1305
          HQ533147.1 - 11 1371
          HQ825316.1 - 11 1374
          HQ831517.1 - 10 1281
          JF500487.1 - 10 1421
          JF500488.1 - 11 1511
          JF500489.1 - 10 1421
          JF500490.1 - 11 1511
          JX220977.1 - 11 1398
          JX254917.1 - 10 1311
          JX268544.1 - 11 1512

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          transport
          ion transport
          sodium ion transport
          anion transport
          regulation of pH
          response to light stimulus
          post-embryonic development
          inorganic anion transport
          bicarbonate transport
          pyramidal neuron development
          regulation of cellular pH
          multicellular organism growth
          locomotory exploration behavior
          sodium ion transmembrane transport
          brain morphogenesis
          regulation of intracellular pH
          anion transmembrane transport
          hydrogen ion transmembrane transport
          transporter activity
          inorganic anion exchanger activity
          anion transmembrane transporter activity
          sodium:bicarbonate symporter activity
          symporter activity
          antiporter activity
          anion:anion antiporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-dhnrf:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0